Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VDA0014

Supelco

Polyethylene (40, 75, 200, 400 mg Cd/kg)

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

manufacturer/tradename

JRC

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

chemicals and industrial polymers

format

neat

SMILES string

C=C

Inchi Key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Packaging

bottles

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoaki Sakata et al.
Transplantation, 87(6), 825-830 (2009-03-21)
There is a recent focus on embolization of the portal vein by transplanted islets as a major cause of early graft loss. The resultant ischemia causes necrosis or apoptosis of cells within the liver. Thus, noninvasive assessment of the liver

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service