Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V2379

Supelco

Veratrine hydrochloride

analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages
forensics and toxicology
veterinary

format

neat

SMILES string

Cl.[H][C@@]12CC[C@H](C)CN1C[C@@]3([H])[C@]4(O)C[C@]56O[C@]7(O)[C@H](CC[C@@]5(C)[C@]7([H])CC[C@@]6([H])[C@]4(O)C[C@H](O)[C@]3(O)[C@]2(C)O)OC(=O)\C(C)=C/C

InChI

1S/C32H49NO9.ClH/c1-6-18(3)25(35)41-24-11-12-26(4)19-8-9-20-28(37)13-23(34)31(39)21(29(28,38)16-30(20,26)42-32(19,24)40)15-33-14-17(2)7-10-22(33)27(31,5)36;/h6,17,19-24,34,36-40H,7-16H2,1-5H3;1H/b18-6-;/t17-,19-,20-,21-,22-,23-,24-,26-,27+,28+,29+,30+,31-,32-;/m0./s1

InChI key

VSARLHXBMYVYSM-ZLVREPKBSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

A mixture of alkaloids, including veratridine, cevadilline, sabadine, and cevadine hydrochlorides.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service