UST102
TPH Surrogate
certified reference material, 6000 μg/mL in methylene chloride: carbon disulfide (1:1)
About This Item
grade
certified reference material
Quality Level
CofA
certificate is enclosed in each package
manufacturer/tradename
RTC UST-102
concentration
6000 μg/mL in methylene chloride: carbon disulfide (1:1)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Analyte
signalword
Danger
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
target_organs
Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
70.3 °F
flash_point_c
21.3 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service