Key Documents
UBBEL16UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 5, Nom. Constant 100.0, Range 20,000 to 100,000 cSt
About This Item
UNSPSC Code:
41116107
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This product is issued with a UKAS 17025 certificate of calibration
Choose from one of the most recent versions:
Certificates of Analysis (COA)