Key Documents
UBBEL12UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 3B, Nom. Constant 5.0, Range 1,000 to 5,000 cSt
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This product is issued with a UKAS 17025 certificate of calibration
Choose from one of the most recent versions:
Certificates of Analysis (COA)