Key Documents
UBBEL05UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 1C, Nom. Constant 0.03, Range 6 to 30 cSt
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified) is used for the determination of kinematic viscosity of transparent and opaque Newtonian liquids.
Features and Benefits
  • High quality glass capillary viscometer.
  • Manufactured in accordance with ASTM D445, D446 and ISO 3104, 3105.
  • Available with a UKAS 17025 certificate of calibration or non-calibrated.
  • Low level of uncertainty of measurement.
Choose from one of the most recent versions:
Certificates of Analysis (COA)