Key Documents
UBBEL02UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 0C, Nom. Constant 0.003, Range 0.6 to 3 cSt
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This product is issued with a UKAS 17025 certificate of calibration
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Abigail S Tyrell et al.
The Biological bulletin, 239(1), 62-71 (2020-08-20)
AbstractChanges in temperature alter the viscosity of fluids, which impacts the force needed to move and the diffusion rates of gases. This is particularly salient for organisms that operate at mid to low Reynolds numbers. In this study, we investigated
Abigail S Tyrell et al.
The Journal of experimental biology, 223(Pt 13) (2020-06-13)
Calanoid copepods, depending on feeding strategy, have different behavioral and biological controls on their movements, thereby responding differently to environmental conditions such as changes in seawater viscosity. To understand how copepod responses to environmental conditions are mediated through physical, physiological