Key Documents
U05NC
BS/U-tube Viscometer for Transparent Liquids
Size E, Nom. Constant 0.3, Range 60 to 300 cSt, (non certified)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
BS/U-tube Viscometer is used for the determination of kinematic viscosity of transparent Newtonian liquids.
Features and Benefits
  • Made from high-quality glass.
  • Capillary viscometer is manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105.
Choose from one of the most recent versions:
Certificates of Analysis (COA)