Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SK017

Supelco

HVOC MATRIX SPIKE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

Quality Level

format

multi-component solution

storage temp.

15-25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    1,2-Dichlorobenzene 40 mg/mL
    Methylene Chloride 120 mg/mL
    1,1,1-Trichloroethane 40 mg/mL

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Ozone 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

19.4 °F

flash_point_c

-7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service