Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6503

Supelco

Silica gel Inorganic Sorbent

high-purity grade, 1000-2500 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:

Product Name

Silica gel, high-purity grade, Type G, 5-15 μm, suitable for thin layer chromatography (TLC)

grade

high-purity grade

Quality Level

type

Type G

form

powder

contains

CaSO4 as binder

technique(s)

LPLC: suitable
thin layer chromatography (TLC): suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

5-15 μm

pore size

60 Å mean pore size

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A M García-Bores et al.
Journal of photochemistry and photobiology. B, Biology, 167, 72-81 (2017-01-01)
Lippia graveolens HBK (Mexican oregano) is a species that is regularly used as a condiment in Mexican cuisine. In traditional medicine, it is used for the treatment of respiratory and digestive illnesses, headaches, rheumatism and inflammation-related disorders. The main chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service