Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5147

Supelco

Secobarbital-d5

analytical standard, 97 atom % D

Synonym(s):

5-(Allyl-d5)-5-(1-methylbutyl)barbituric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13D5N2O3
CAS Number:
Molecular Weight:
243.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

isotopic purity

97 atom % D

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[Na].[H]\C([H])=C(\[H])C([H])([H])C1(C(C)CCC)C(=O)NC(=O)NC1=O

InChI

1S/C12H18N2O3.Na.H/c1-4-6-8(3)12(7-5-2)9(15)13-11(17)14-10(12)16;;/h5,8H,2,4,6-7H2,1,3H3,(H2,13,14,15,16,17);;

InChI key

DAOMPCXXNPYORA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service