S2400006
Suxibuzone impurity C
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Butyl-4-(hydroxymethyl)-1,2-diphenyl-1,2-dihydro-4H-pyrazole-3,5-dione, 4-Hydroxymethylphenylbutazone
Select a Size
$118.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$118.00
Notify Me
Get notified when this item is ready to ship via email.
Recommended Products
grade
pharmaceutical primary standard
API family
suxibuzone
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N2(N(C(=O)C(C2=O)(CO)CCCC)c3ccccc3)c1ccccc1
InChI
1S/C20H22N2O3/c1-2-3-14-20(15-23)18(24)21(16-10-6-4-7-11-16)22(19(20)25)17-12-8-5-9-13-17/h4-13,23H,2-3,14-15H2,1H3
InChI key
VCUPPSLLQJDTJC-UHFFFAOYSA-N
Related Categories
General description
Application
Packaging
Other Notes
related product
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service