Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S115

Supelco

Nor-S-(−)-SCH-23388 hydrochloride

analytical standard

Synonym(s):

(S)-(−)-7-Chloro-8-hydroxy-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$191.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C16H16ClNO · HCl
CAS Number:
Molecular Weight:
310.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$191.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.Oc1cc2[C@@H](CNCCc2cc1Cl)c3ccccc3

InChI

1S/C16H16ClNO.ClH/c17-15-8-12-6-7-18-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,18-19H,6-7,10H2;1H/t14-;/m0./s1

InChI key

KTHOTRUZLPTFOY-UQKRIMTDSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service