Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RQC03006

Supelco

Salmonella enterica subsp. enterica serovar Abaetetuba 500 CFU

certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

certified reference material

Quality Level

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Salmonella spp.

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service