Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RQC02351

Supelco

Clostridium perfringens Vitroids 30 CFU

30 CFU, certified reference material, suitable for microbiology

Synonym(s):

Clostridium perfringens Vitroids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Product RQC02351 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

manufacturer/tradename

RTC RQC-0235-30

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Clostridium spp.

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Legal Information

Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service