Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RQC017550

Escherichia coli O157:H7 ATCC® 43895 RapidCheck 50 CFU

certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
Pricing and availability is not currently available.

grade

certified reference material

manufacturer/tradename

RTC RQC-0175-50

matrix

Vitroid

application(s)

environmental
food and beverages

microbiology

suitability

Escherichia coli
coliforms

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 (ISO Guide 34) and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot.

Legal Information

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative Word mark, and the ATCC Catalog Marks are trademarks of ATCC. RTC Sigma, Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
ATCC is a registered trademark of American Type Culture Collection
RapidCheck is a trademark of RT Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service