Key Documents
RF11NC
BS/IP/RF U-Tube Reverse Flow Viscometer for Transparent and Opaque Liquids
(Size 11, Nom.Constant 300, Range 60,000 to 300,000 cSt)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Choose from one of the most recent versions:
Certificates of Analysis (COA)