Key Documents
RF01UKC
BS/IP/RF U-Tube Reverse Flow Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Size 1, Nom.Constant 0.003, Range 0.6 to 3 cSt
About This Item
UNSPSC Code:
41116107
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This product is issued with a UKAS 17025 certificate of calibration
Choose from one of the most recent versions:
Certificates of Analysis (COA)