Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2253

Supelco

Rolitetracycline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33N3O8
CAS Number:
Molecular Weight:
527.57
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[H][C@@]12C[C@@]3([H])C(=C(O)[C@]1(O)C(=O)C(C(=O)NCN4CCCC4)=C(O)[C@H]2N(C)C)C(=O)c5c(O)cccc5[C@@]3(C)O

InChI

1S/C27H33N3O8/c1-26(37)13-7-6-8-16(31)17(13)21(32)18-14(26)11-15-20(29(2)3)22(33)19(24(35)27(15,38)23(18)34)25(36)28-12-30-9-4-5-10-30/h6-8,14-15,20,31,33-34,37-38H,4-5,9-12H2,1-3H3,(H,28,36)/t14-,15-,20-,26+,27-/m0/s1

InChI key

HMEYVGGHISAPJR-IAHYZSEUSA-N

General description

Chemical structure: tetracycline

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Rencken et al.
The Journal of urology, 143(5), 940-943 (1990-05-01)
Sclerotherapy with 3% sodium tetradecyl sulfate and 3.5% rolitetracycline on an outpatient basis was applied to 55 hydroceles. The over-all cure rate was 96% with an average followup of 13 months. Of the patients 64% were cured after only 1
M Carrozzo et al.
Minerva medica, 78(4), 243-246 (1987-02-28)
A new and easy technique for the fluorimetric evaluation of rolitetracycline calcium complex was studied. The study was carried out in 3 different groups of rats; an increase in power of fluorescence in the rats on a low calcium diet
M Hirsch et al.
Sbornik vedeckych praci Lekarske fakulty Karlovy univerzity v Hradci Kralove. Supplementum, 33(5), 721-729 (1990-01-01)
Authors studied the oxygen demand by E. coli suspensions (serotypes 023 and 026) modifying it with various concentration of Rolitetracyclin, and proceeded by comparison of both serotypes. Time dependence of oxygen demand has been determined using the direct Warburg's manometric
K Yokoyama
[Hokkaido igaku zasshi] The Hokkaido journal of medical science, 61(3), 379-387 (1986-05-01)
The purpose of the present study was to investigate the effect of Tetracyclines (TCs); Minocycline (MINO), Tetracycline (TC), Demethylchlortetracycline (DMTC), Pyrrolidinomethyltetracycline (PMTC), on amino acid transport and incorporation in hepatocytes. By the use of liver cell culture, this study was
F W Jennings
Research in veterinary science, 43(2), 173-176 (1987-09-01)
The simultaneous administration, to Trypanosoma brucei infected mice, of rolitetracycline or oxytetracycline and diminazene aceturate appeared to have no effect on the initial trypanocidal action of the diminazene aceturate in that trypanosomes were cleared from the circulation. It also had

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service