Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R0773

Millipore

Rappaport Vassiliadis medium

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

5.2±0.2 (25 °C)

application(s)

environmental
food and beverages
sample preparation

microbiology

suitability

selective for Salmonella spp.

Application

Recommended for enrichment of Salmonellae.

Components

Components (g/L)
Papaic digest of soybean meal 4.50
Sodium chloride 7.20
Monopotassium phosphate 1.44
Magnesium chloride 36.00
Malachite Green 0.036
Yeast extract 2.50
Dextrose 1.00
Agar 9.00

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M van Schothorst et al.
The Journal of applied bacteriology, 54(2), 209-215 (1983-04-01)
Enhanced growth of salmonellas in Rappaport's medium as modified by Vassiliadis et al. (1976) after pre-enrichment in buffered peptone water during the first 6 h was obtained by replacement of tryptone by soya peptone. The competing bacteria, i.e. those which
Van Schothorst, M., et al.
Food Microbiology, 4, 11-11 (1987)

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service