QCMIC211
Streptococcus/Enterococcus - Drinking & Surface Water QC 4 Levels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106200
grade
certified reference material
packaging
set of
manufacturer/tradename
RTC QC-MIC-211
application(s)
environmental
food and beverages
shipped in
wet ice
storage temp.
2-8°C
suitability
Enterococcus spp.
Streptococcus spp.
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Packaging
Set contains 8 sterile screw cap vials each containing a Vitroid (a small disk containing the organisms) and 8 - 99mL bottles of phosphate buffer.
Other Notes
Refer to the Certificate of Analysis for additional information.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service