Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QCMIC014

E. coli - Sludge QC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

certified reference material

packaging

set of

manufacturer/tradename

RTC QC-MIC-014

application(s)

environmental
food and beverages

shipped in

wet ice

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Packaging

This sample is provided as a stable Vitroid requiring hydration. The sample is provided in duplicate. A 50g of sterile sludge is supplied with the sample. This product does not contain the phosphate buffered hydrolyzing solution. The buffer can be ordered separately under the number Z699489(Aldrich).

Other Notes

This sample is designed for the quantitative analysis of fecal coliform and e coli. Refer to the Certificate of Analysis for additional information.

Legal Information

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service