Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC3041

Supelco

Minerals - Whole Volume

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-3027-12

concentration

in H2O

technique(s)

AAS: suitable
ICP: suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Preparation Note

This sample is packaged in a 500 mL HDPE bottle containing approximately 500 mL of solution.
This sample is not preserved.


Sample preparation
1) This sample has been prepared as a whole volume sample and is ready to be analyzed as received.
2) Shake bottle well prior to use.
3) Analyze alkalinity first. Be sure to recap the bottle when not in use to minimize CO2 absorption.
4) Sample and analyze using your normal methods.
5) Analyze within 28 day hold time.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Alkalinity as CaCO3 25 - 400 mg/L
    Calcium hardness as CaCO3 25 - 250 mg/L
    Calcium, Ca 10 - 100 mg/L
    Hardness 17- 440 mg/L
    Hardness, total as CaCO3 40 - 415 mg/L
    Magnesium, Mg 4 - 40 mg/L
    Potassium, K 4 - 40 mg/L
    Sodium, Na 10 - 100 mg/L
    See All (8)

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service