Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1453

Supelco

Chromium VI - WS

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

packaging

ampule of 20 mL

manufacturer/tradename

RTC QCI-229

concentration

100% in H2O

technique(s)

AAS: suitable
ICP: suitable

application(s)

environmental
food and beverages

format

matrix material

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Pytlakowska et al.
Mikrochimica acta, 187(9), 542-542 (2020-09-04)
Molybdenum disulfide (MoS2) was supported on graphene oxide (GO) by hydrothermal method. The resulting nanocomposite (MoS2-rGO) was characterized by X-ray photoelectron spectroscopy, scanning electron microscopy, and transmission electron microscopy. The experiments show that at pH 2, MoS2-rGO has a great

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service