Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1274

Supelco

Acids - WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product QC1274 is not currently sold in your country. Contact Technical Service

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 2 mL

manufacturer/tradename

RTC QCO-022

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Benzoic acid
    4-Chloro-3-methylphenol
    2-Chlorophenol
    2,4-Dichlorophenol
    2,6-Dichlorophenol
    2,4-Dimethylphenol
    2-Methyl-4,6-dinitrophenol
    o-Cresol
    m-Cresol
    p-Cresol
    2-Nitrophenol
    Pentachlorophenol
    Phenol
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    See All (15)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

1.4 °F - closed cup - Solvent

flash_point_c

-17 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service