Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1253

Supelco

Chlorinated hydrocarbons

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

storage temp.

2-30°C

Analyte

Description

    1,2,4-Trichlorobenzene 1.00 - 200µg/L
    1,2-Dichlorobenzene 1.00 - 200µg/L
    1,3-Dichlorobenzene 1.00 - 200µg/L
    1,4-Dichlorobenzene 1.00 - 200µg/L
    2-Chloronaphthalene 1.00 - 200µg/L
    Hexachlorobenzene 1.00 - 200µg/L
    Hexachlorobutadiene 1.00 - 200µg/L
    Hexachlorocyclopentadiene 1.00 - 200µg/L
    Hexachloroethane 1.00 - 200µg/L
    See All (9)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service