Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1223

Supelco

Low Level PAHs - WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 2 mL

manufacturer/tradename

RTC QCO-259

concentration

100% in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Naphthalene
    Acenaphthene
    Acenaphthylene
    Anthracene
    Benzo[a]pyrene
    Benzo[b]fluoranthene
    Benzo[ghi]perylene
    Benzo[k]fluoranthene
    Chrysene
    Dibenz[a,h]anthracene
    Fluoranthene
    Fluorene
    Indeno[1,2,3-c,d]pyrene solution
    Phenanthrene
    Pyrene
    See All (15)

pictograms

FlameExclamation mark

signalword

Danger

supp_hazards

flash_point_f

Solvent

flash_point_c

Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krystyna Kozioł et al.
The Science of the total environment, 596-597, 158-168 (2017-04-23)
Glaciers accumulate organic pollutants delivered by snow. However, our understanding of the exact dynamics of organic pollutants in the snowpack relies primarily on laboratory experiments and mathematical models. To fill the gap related to the detailed field data, we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service