Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1209

Supelco

Uranium - WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 20 mL

manufacturer/tradename

RTC QCI-264

technique(s)

AAS: suitable
ICP: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service