Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1121

Supelco

pH 4 Buffer

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-027-4-0.100

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

These products are discontinued but the list below shows products that can be used as replacement.

Obsolete Part NumberReplacement Part Number
QC1121-100ML1990640001
QC1121-250ML1094450500
QC1121-500ML1094450500
QC1121-1L1094061000
QC1121-10L1094359010

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service