QC1011
Nitrogen Pesticides
certified reference material
About This Item
grade
certified reference material
CofA
current certificate can be downloaded
manufacturer/tradename
RTC QCO-122-3
concentration
in acetone
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
matrix material
storage temp.
2-8°C
General description
Application
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
1.4 °F - closed cup - Solvent
flash_point_c
-17 °C - closed cup - Solvent
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service