Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR2516

Supelco

Naproxen Related Compound G

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (2R)-2-(6-Methoxy-2-naphthyl)propionic acid, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
pharmaceutical secondary standard

agency

traceable to USP 1457436

API family

naproxen

packaging

pkg of 100 mg

mp

156-158 °C (lit.)

storage temp.

2-30°C

SMILES string

COc1ccc2cc(ccc2c1)[C@@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m1/s1

InChI key

CMWTZPSULFXXJA-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4567 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service