Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1495

Supelco

Trientine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Triethylenetetramine dihydrochloride, N,N′-Bis(2-aminoethyl)-1,2-ethanediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2 · 2HCl
CAS Number:
Molecular Weight:
219.16
Beilstein/REAXYS Number:
5961763
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

agency

traceable to USP 1683504

API family

trientine hydrochloride

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.NCCNCCNCCN

InChI

1S/C6H18N4.2ClH/c7-1-3-9-5-6-10-4-2-8;;/h9-10H,1-8H2;2*1H

InChI key

WYHIICXRPHEJKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Trientine Hydrochloride is administered for relief of symptoms of Wilson′s disease for penicillamine intolerance, hence is employed as an oral medication for the treatment of an inherited defect in copper metabolism.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3043 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ARABIC-Mosby's Pocket Dictionary of Medicine, Nursing & Health Professions (2009)
ARABIC-Mosby's Pocket Dictionary of Medicine, Nursing & Health Professions (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service