Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1453

Supelco

Erythrulose

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

L-(+)-Erythrulose, S-1,3,4-Trihydroxy-2-butanone, L-Glycero-2-tetrulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC[C@H](O)C(=O)CO

InChI

1S/C4H8O4/c5-1-3(7)4(8)2-6/h3,5-7H,1-2H2/t3-/m0/s1

InChI key

UQPHVQVXLPRNCX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Erythrulose is generally used as a tanning agent in the cosmetics industry and is a source of chiral ethyl ketones used in aldol reaction organic synthesis. Red raspberries are a natural source of erythrulose. It is often used in combination of dihydroxyacetone to impart attractive tanning properties to the skin.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3001 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Trassaert et al.
Microbial cell factories, 16(1), 141-141 (2017-08-16)
The oleaginous yeast Yarrowia lipolytica is increasingly used as alternative cell factory for the production of recombinant proteins. At present, several promoters with different strengths have been developed based either on the constitutive pTEF promoter or on oleic acid inducible
Xia Hua et al.
Biotechnology for biofuels, 13, 1-1 (2020-01-09)
The critical issue in the competitiveness between bioengineering and chemical engineering is the products titer and the volume productivity. The most direct and effective approach usually employs high-density biocatalyst, while the weakened mass transfer and evoked foam problem accompany ultrahigh-density
A review of common tanning methods
Garone M, et al.
The Journal of Clinical and Aesthetic Dermatology, 8(2), 43-43 (2015)
Chrispian W Theron et al.
Scientific reports, 10(1), 1741-1741 (2020-02-06)
The large-scale production of recombinant proteins (rProt) is becoming increasingly economically important. Among the different hosts used for rProt production, yeasts are gaining popularity. The so-called non-conventional yeasts, such as the methylotrophic Pichia pastoris and the dimorphic Yarrowia lipolytica, are
Henry C Wilkinson et al.
Scientific reports, 10(1), 4148-4148 (2020-03-07)
We recently characterised a low-activity form of E. coli transketolase, TKlow, which also binds the cofactor thiamine pyrophosphate (TPP) with an affinity up to two-orders of magnitude lower than the previously known high TPP-affinity and high-activity form, TKhigh, in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service