Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PFS62

Supelco

SpheriCal Mix CHCA

Peptide Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

SpheriCal

form

powder

shelf life

expiry date on the label

composition

α-cyano-4-hydroxycinnamic acid, >90%
sodium trifluoroacetate, <10%

Storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibration spots, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i.SpheriCal Mix DHB Peptide Low(m/z 500 - 1,600)
ii.SpheriCal Mix DHB Peptide Medium(m/z 1,600 - 3,500)
iii.SpheriCal Mix DHB Peptide High(m/z 3,500 - 7,500)
iv.SpheriCal Mix DHB Protein Low (m/z 7,500 - 15,000)
v.SpheriCal DHB Protein Medium (m/z 15,000 - 30,000)

SpheriCal Mix: quick and easy to use - just add solvent

As calibration standard, matrix and counter ion are pre-mixed in optimized ratios, SpheriCal Mix dramatically expedites sample preparation for calibration in minutes. SpheriCal Mix CHCA is pre-formulated with α-cyano-4-hydroxycinnamic acid, a commonly used matrix in the analysis of peptides, proteins, phospholipids, nucleotides and polar synthetic polymers.

Find more information here SpheriCal

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service