Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PFS43

Supelco

SpheriCal Mix DCTB

Protein Low

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

shelf life

expiry date on the label

composition

trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile (DCTB), >80%
sodium trifluoroacetate, <20%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

calibrants PFS4A : PFS4B : PFS4C : PFS4D = 1:1:1:1

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service