Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PFS36

Supelco

SpheriCal Aqua Mix DHB

Peptide Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

shelf life

expiry date on the label

composition

2,5-dihydroxybenzoic acid (DHB), >90%
sodium trifluoroacetate, <10%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibrations, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i.SpheriCal Aqua Mix Peptide Low (m/z 500 - 1,000)
ii.SpheriCal Aqua Mix Peptide Medium (m/z 1,000 - 2,500)
iii.SpheriCal Aqua Mix Peptide High (m/z 2,500 - 5,200)
iv.SpheriCal Mix Protein Low (m/z 5,200 - 11,000)

SpheriCal Aqua Neat: ideal for water-based systems.

SpheriCal Aqua Mix is ideal for calibration with aqueous systems commonly used in life sciences. As calibration standard, matrix and counter ion are pre-mixed in optimized ratios, SpheriCal Aqua Mix dramatically expedites sample preparation for calibration in minutes. SpheriCal Mix is pre-formulated with DHB, a commonly used matrix in the analysis of peptides, proteins, phospholipids, nucleotides and polar synthetic polymers. Each vial of SpheriCal Aqua Mix contains sufficient calibration standard for over 100 calibration spots.

Find more information here SpheriCal
dendritic calibrants PFQ2A : PFQ2B : PFQ2C : PFQ2D = 1:1:1:1

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service