PFS24
SpheriCal™ Neat
Protein Medium, suitable for MALDI-MS
About This Item
product line
SpheriCal™
form
solid
shelf life
expiry date on the label
mol wt
Mr 15000-30000 Da (Na & adduct)
technique(s)
MALDI-MS: suitable
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Mass ranges available:
i.SpheriCal™ Neat Peptide Low (m/z 500 - 1,600)
ii.SpheriCal™ Neat Peptide Medium (m/z 1,600 - 3,500)
iii.SpheriCal™ Neat Peptide High (m/z 3,500 - 7,500)
iv.SpheriCal™ Neat Protein Low (m/z 7,500 - 15,000)
v.SpheriCal™ Neat Protein Medium(m/z 15,000 - 30,000).
SpheriCal™ Neat: unprecedented freedom for the user.
SpheriCal™ Neat is provided without any matrix or added cation and, because the SpheriCal™ calibration standards exhibit unusually broad solvent and matrix compatibility, this product can be used with a wide range of sample preparation conditions, though ionization is preferred with alkali cations. The freedom of this product line will enable you to explore your own calibration protocols for optimization purposes. Each vial of SpheriCal™ Neat contains sufficient calibration standard for over 100 calibration spots.
Find more information here SpheriCal™
Application
Packaging
Legal Information
related product
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service