Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PEA1924

Supelco

Pesticides on PUF

Proficiency Testing Material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

agency

EPA (Method TO-4, TO-10A Organo Chlorine Pesticides on PUF)

packaging

package of 1 ea

manufacturer/tradename

RTC A-1924

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

The sample consists of one PUF (polyurethane foam) plug that has been spiked with Pesticides.

Sample preparation
Remove plug from the 20 mL VOA vial.  Use caution, free liquid may be present.  
Place the PUF plug into the extraction glassware to be used.  
Rinse the VOA vial that contained the PUF plug with the extraction solvent to be used at least three times to transfer the entire contents of the vial.  
Extract the sample using your normal procedures.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Air
AIR20-2 April 8 - May 22, 2020
AIR20-4 October 7 - November 20, 2020

AIR21-2 April 7 - May 21, 2021
AIR21-4 October 6 - November 19, 2021

Quick-Turn Study Not Available for this Product

Analyte

Description

    Aldrin 1-10 μg/PUF
    δ-BHC 1-10 μg/PUF
    α-BHC (alpha-Hexachlorocyclohexane) 1-10 μg/PUF
    β-BHC (beta-Hexachlorocyclohexane) 1-10 μg/PUF
    γ-BHC (Lindane, gamma-Hexachlorocyclohexane) 1-10 μg/PUF
    α-Chlordane 1-10 μg/PUF
    γ-Chlordane 1-10 μg/PUF
    4,4'-DDD 1-10 μg/PUF
    4,4′-DDE 1-10 μg/PUF
    4,4′-DDT 1-10 μg/PUF
    Dieldrin 1-10 μg/PUF
    Endosulfan I 1-10 μg/PUF
    Endosulfan II 1-10 μg/PUF
    Endosulfan sulfate 1-10 μg/PUF
    Endrin aldehyde 1-10 μg/PUF
    See All (15)

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service