Certificates of Analysis are not available for RTC proficiency testing products. Those are the samples with the SPE or PE prefix in the part number.
packaging
ampule of 1 × 2 mL
manufacturer/tradename
RTC PEO-115
concentration
in acetone
application(s)
environmental
storage temp.
2-8°C
General description
Preparation Note
TPH consists of a light petroleum hydrocarbon (>12 to <35).
The solvent is Freon 113. This solvent has the proper boiling point and IR spectra as to not interfere with the methods.
Handle the concentrate in a properly ventilated hood.
Sample preparation
Transfer exactly 1 mL of concentrate directly to the separatory funnel.
Add 1 L of reagent grade DI water to the separatory funnel.
Considered this as your sample for test.
Proceed with analysis.
TPH is reported in mg/L. Assume sample volume is 1-Liter for calculation purposes.
Other Notes
Proficiency Testing Schedule (2016-2017)
Wastewater
WP16-1January 13 - February 26, 2016
WP16-2March 23 - May 6, 2016
WP16-3AMay 11 - June 24, 2016
WP16-3BJuly 6 - August 19, 2016
WP16-4September 21 - November 4, 2016
WP16-5November 23 - January 6, 2017
WP17-1January 18 - March 3, 2017
WP17-2March 29 - May 12, 2017
WP17-3AMay 17 - June 30, 2017
WP17-3BJuly 12 - August 25, 2017
WP17-4September 27 - November 10, 2017
WP17-5November 22 - January 5, 2018
Quick-Turn Study - (non-scheduled proficiency test that is available at any time)
Analyte
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Muta. 1B - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-56.2 °F - closed cup - Solvent
flash_point_c
-49 °C - closed cup - Solvent
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Are Certificates of Analysis created for RTC proficiency testing products?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service