Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PE1285

Supelco

Sulfur in Water - WP

Proficiency Testing Material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

ampule of 1 × 20 mL

manufacturer/tradename

RTC PEI-259

concentration

in H2O

application(s)

environmental

format

matrix material

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is packaged as a 20mL concentrate.
The solvent for this sample is water.
The PT sample has been autoclaved for preservation

Sample Preparation
Partially fill a 1000mL volumetric flask with reagent-grade water (near 20ºC).
Open ampule PE1285-20ML (be careful to avoid cuts on glass) and transfer 10.0mL of concentrate to the volumetric flask.
Fill the volumetric flask to volume with reagent-grade water and mix well.
The sample is now ready for immediate analysis.
Sealed ampules were autoclaved to preserve the solutions inside. This preservation treatment is not effective after the ampule is opened. Therefore, the sample should be analyzed as soon as possible after sample preparation.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2019-2020)

Wastewater
WP19-1January 16 - March 1, 2019
WP19-2March 27 - May 10, 2019
WP19-3AMay 15 - June 28, 2019
WP19-3BJuly 10 - August 23, 2019
WP19-4September 25 - November 8, 2019
WP19-5November 20 - January 3, 2020

WP20-1January 15 - February 28, 2020
WP20-2March 25 - May 8, 2020
WP20-3AMay 13 - June 26, 2020
WP20-3BJuly 8 - August 21, 2020
WP20-4September 23 - November 6, 2020
WP20-5November 25 - January 8, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

    Sulfur, S 1-100 mg/L

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service