Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PE1151

Supelco

Herbicides 2A - WP

Proficiency Testing Material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

manufacturer/tradename

RTC PEO-094-2A

concentration

in tert-butyl methyl ether

application(s)

agriculture
environmental

format

matrix material

storage temp.

2-8°C

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is packaged as a 1.5 mL concentrate.
The solvent for this sample is MTBE.
The PT sample is not preserved.

Sample Preparation
To prepare solution, partially fill a 1000 mL volumetric flask with reagent grade water.
Open ampule PEO-094-2A and transfer exactly 1.0 mL to the volumetric flask.
Fill the flask to volume and mix well.
This represents the sample ready for analysis.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2019-2020)

Wastewater
WP19-1January 16 - March 1, 2019
WP19-2March 27 - May 10, 2019
WP19-3AMay 15 - June 28, 2019
WP19-3BJuly 10 - August 23, 2019
WP19-4September 25 - November 8, 2019
WP19-5November 20 - January 3, 2020

WP20-1January 15 - February 28, 2020
WP20-2March 25 - May 8, 2020
WP20-3AMay 13 - June 26, 2020
WP20-3BJuly 8 - August 21, 2020
WP20-4September 23 - November 6, 2020
WP20-5November 25 - January 8, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

    Asulam 30-150 ng/L
    Carbendazim 30-150 ng/L

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-27.4 °F - closed cup - Solvent

flash_point_c

-33 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service