Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PE1075

Supelco

Speciation-arsenic and selenium - Proficiency Testing (PT)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

description

Proficiency Testing Material

concentration

in H2O

format

matrix material

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

Description
1) This PT sample is packaged as a 20 mL concentrate.
2) The solvent for this sample is DI water.
3) The PT sample has not been preserved.

Sample Preparation PLEASE REFER TO YOUR INSTRUCTION PACKET FOR LATEST INSTRUCTIONS
1) To prepare solution partially fill a 1000-mL volumetric flask with reagent-grade water (near 20 degrees C).
2) Shake ampule well prior to opening.
3) Open ampule PECS-1073 (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask.
4) Fill the flask to volume with reagent-grade water and mix well.
5) The sample is now ready for immediate analysis.
6) This represents the sample for analysis.
7) Report rssults in mg/L.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2018-2019)

Wastewater
WP18-1January 17 - March 2, 2018
WP18-2March 28 - May 11, 2018
WP18-3AMay 16 - June 29, 2018
WP18-3BJuly 11 - August 24, 2018
WP18-4September 26 - November 9, 2018
WP18-5November 21 - January 4, 2019

WP19-1January 16 - March 1, 2019
WP19-2March 27 - May 10, 2019
WP19-3AMay 15 - June 28, 2019
WP19-3BJuly 10 - August 23, 2019
WP19-4September 25 - November 8, 2019
WP19-5November 20 - January 3, 2020

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

    Arsenic (III) 500-1500 ug/L
    Arsenic (V) 500-1500 ug/L
    Selenium (IV) 500-1500 ug/L
    Selenium (VI) 500-1500 ug/L
    Selenium 50-2000 ug/L

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service