Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8369

Supelco

Protein standard solution

analytical standard, 0.5 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701

grade

analytical standard

Quality Level

analyte chemical class(es)

amino acids, peptides, proteins

concentration

0.5 mg/mL

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

Other Notes

Human serum albumin in saline containing 0.1% sodium azide as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeoun Jin Kim et al.
Nature protocols, 7(5), 859-871 (2012-04-14)
Mass spectrometry-based targeted proteomics is a rapidly expanding method for quantifying proteins in complex clinical samples such as plasma. In conjunction with the stable isotope dilution method, selected reaction monitoring (SRM) assays provide unparalleled sensitivity and selectivity for detection and
Krzysztof Wrzesinski et al.
Methods in molecular biology (Clifton, N.J.), 2273, 17-62 (2021-02-20)
Growing cells as 3D structures need not be difficult. Often, it is not necessary to change cell type, additives or growth media used. All that needs to be changed is the geometry: cells (whether primary, induced pluripotent, transformed or immortal)

Related Content

Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service