P5592
Potassium peroxodisulfate
BioXtra, ≥99.0%
Synonym(s):
Potassium persulfate
About This Item
Recommended Products
product line
BioXtra
assay
≥99.0%
reaction suitability
reagent type: oxidant
impurities
≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter
solubility
H2O: 0.5 M, clear, colorless
anion traces
chloride (Cl-): ≤0.001%
cation traces
Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.01%
NH4+: ≤0.5%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%
SMILES string
[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
InChI
1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
InChI key
USHAGKDGDHPEEY-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
5.1B - Oxidizing hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service