Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3959

Sigma-Aldrich

Potassium iodate

BioXtra, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioXtra

assay

≥99.0%

impurities

<0.005% Insoluble matter
<0.01% Phosphorus (P)

mp

560 °C (lit.)

solubility

H2O: 0.1 M at 25 °C, clear, colorless

density

3.93 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): <0.005%

cation traces

Al: <0.0005%
Ca: <0.01%
Cu: <0.0005%
Fe: <0.0005%
Mg: <0.005%
NH4+: <0.05%
Na: <0.5%
Pb: <0.001%
Zn: <0.0005%

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Rohner et al.
Food and nutrition bulletin, 33(4 Suppl), S330-S335 (2013-03-01)
Despite considerable progress made in the past decade through salt iodization programs, over 2 billion people worldwide still have inadequate iodine intake, with devastating consequences for brain development and intellectual capacity. To optimize these programs with regard to salt iodine
Ralf Biebinger et al.
The British journal of nutrition, 102(9), 1362-1369 (2009-08-06)
Adverse sensory changes prevent the addition of highly bioavailable ferrous sulfate (FeSO4) to most wheat flours. Poorly absorbable reduced Fe powders are commonly used. Encapsulation of FeSO4 can overcome these sensory changes, but the particle size of commercial compounds is
Gehad G Mohamed et al.
Drug testing and analysis, 2(1), 28-36 (2010-09-30)
Two spectrophotometric procedures are suggested for the determination of three irreversible proton pump inhibitors, rabeprazole (RAB), omeprazole (OMP) and pantoprazole (PAN) in pure form and in different pharmaceutical formulations. The first method is based on the oxidation of RAB and
Wim Voogt et al.
Journal of the science of food and agriculture, 90(5), 906-913 (2010-04-01)
Iodine is an essential trace element for humans. Two billion individuals have insufficient iodine intake. Biofortification of vegetables with iodine offers an excellent opportunity to increase iodine intake by humans. The main aim was to study the effect of iodine
Xue F Yang et al.
The British journal of nutrition, 98(1), 116-122 (2007-04-26)
Excessive iodine induces thyroid dysfunction. However, the effect of excessive iodine exposure on maternal-fetal thyroid hormone metabolism and on the expression of genes involved in differentiation, growth and development is poorly understood. Since a thyroid hormone receptor response element was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service