Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3421

Supelco

5α-Pregnane-3,11,20-trione

analytical standard, for drug analysis

Synonym(s):

Allopregnanetrione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥90%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(C)=O)[C@@]4(C)CC(=O)[C@]3([H])[C@@]1(C)CCC(=O)C2

InChI

1S/C21H30O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h13,15-17,19H,4-11H2,1-3H3/t13-,15-,16+,17-,19+,20-,21+/m0/s1

InChI key

AHRWWYGWQKBKBF-MUGXHADPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachelle Gent et al.
The Journal of steroid biochemistry and molecular biology, 189, 116-126 (2019-03-03)
The 11β-hydroxysteroid dehydrogenase (11βHSD) types 1 and 2 are primarily associated with glucocorticoid inactivation and reactivation. Several adrenal C11-oxy C19 and C11-oxy C21 steroids, which have been identified in prostate cancer, 21-hydroxylase deficiency and polycystic ovary syndrome, are substrates for
Desmaré van Rooyen et al.
The Journal of steroid biochemistry and molecular biology, 199, 105614-105614 (2020-02-03)
Cytochrome P450 17α-hydroxylase/17,20-lyase (CYP17A1) plays a pivotal role in the regulation of adrenal and gonadal steroid hormone biosynthesis. More recent studies highlighted the enzyme's role in the backdoor pathway leading to androgen production. Increased CYP17A1 activity in endocrine disorders and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service