Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0727

Millipore

Phenol red starch broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder

application(s)

microbiology

Application

Recommended for studying the fermentation of starch by microorganisms.

Components

Ingredients (g/L)
Proteose peptone, 10.00
Beef extract, 1.00
Sodium chloride, 5.00
Starch, 5.00
Phenol red, 0.018

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunyao Xu et al.
Cell communication and signaling : CCS, 18(1), 104-104 (2020-07-10)
Sepsis is an infection-induced aggressive and life-threatening organ dysfunction with high morbidity and mortality worldwide. Infection-associated inflammation and coagulation promote the progression of adverse outcomes in sepsis. Here, we report that phospho-Tyr705 of STAT3 (pY-STAT3), not total STAT3, contributes to
MacFaddin, J
Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria, 1 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service