Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0556

Millipore

Bacteriological Peptone

Enzymatic hydrolysate

Synonym(s):

Peptone from animal tissue

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106200

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For the preparation of general culture media.

Quality

Bacteriological peptone contains 14.2 - 17.4% total nitrogen in a readily available form for bacteria. It also has a high peptide and amino acid content.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyadharshini Ramachandran et al.
Journal of microbiology and biotechnology, 23(3), 351-356 (2013-03-07)
A high beta-glucosidase (BGL)-producing strain, Stereum hirsutum, was identified and isolated and showed a maximum BGL activity (10.4 U/ml) when cultured with Avicel and tryptone as the carbon and nitrogen sources, respectively. In comparison with other BGLs, BGL obtained from
Miguel Betancourt-Sanchez et al.
Microbiology (Reading, England), 155(Pt 9), 2895-2906 (2009-06-23)
In an in vitro model using HEp-2 cells treated with purified plasmid-encoded toxin (Pet), we have identified morphological changes characterized by cell rounding and detachment after toxin internalization; these changes progress to cell death. However, these effects have not yet
H B Nielsen et al.
Environmental technology, 28(8), 905-914 (2007-09-21)
Two thermophilic continuously stirred tank reactors, R1 and R2, were subject to pulses of tryptone and ammonia. R1 was operated at an ammonia-N concentration of 3.0 g l(-1) and R2 was operated at an ammonia-N concentration of 1.7 g l(-1).
Eliane Frimmersdorf et al.
Environmental microbiology, 12(6), 1734-1747 (2010-06-18)
In addition to transcriptome and proteome studies, metabolome analysis represents a third complementary approach to identify metabolic pathways and adaptation processes. In order to elucidate basic principles of metabolic versatility of Pseudomonas aeruginosa, we investigated the metabolome profiles of two
Seok Hoon Hong et al.
Nature communications, 3, 613-613 (2012-01-05)
To utilize biofilms for chemical transformations in biorefineries they need to be controlled and replaced. Previously, we engineered the global regulator Hha and cyclic diguanylate-binding BdcA to create proteins that enable biofilm dispersal. Here we report a biofilm circuit that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service