Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O2628

Supelco

Oxycodone hydrochloride solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4 · HCl
CAS Number:
Molecular Weight:
351.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.COc1ccc2C[C@H]3N(C)CC[C@@]45C(Oc1c24)C(=O)CC[C@@]35O

InChI

1S/C18H21NO4.ClH/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10;/h3-4,13,16,21H,5-9H2,1-2H3;1H/t13-,16+,17+,18-;/m1./s1

InChI key

MUZQPDBAOYKNLO-RKXJKUSZSA-N

Gene Information

human ... OPRM1(4988)

Application

Oxycodone hydrochloride may be used as an analytical standard for the determination of oxycodone in plasma samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanogram level quantitation of oxycodone in human plasma by capillary gas chromatography using nitrogen?phosphorus selective detection
Kapil.PR, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 577(2), 283-287 (1992)
Self-assembly of thermally responsive nanoparticles of a genetically encoded peptide polymer by drug conjugation.
Jonathan R McDaniel et al.
Angewandte Chemie (International ed. in English), 52(6), 1683-1687 (2013-01-03)
M Soledad Cepeda et al.
The journal of pain : official journal of the American Pain Society, 14(2), 158-164 (2012-12-21)
Obtaining opioids from multiple prescribers, known as doctor shopping, is 1 example of opioid abuse and diversion. The dual mechanism of action of tapentadol could make tapentadol less likely to be abused than other opioids. The aim of this retrospective
Mari A Kaunisto et al.
Anesthesiology, 119(6), 1410-1421 (2013-12-18)
This article describes the methods and results of the early part (experimental pain tests and postoperative analgesia) of a study that assesses genetic and other factors related to acute pain and persistent pain after treatment of breast cancer in a
Jessica L Asay et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 31(7), 1007-1012 (2013-03-20)
Gait measures are receiving increased attention in the evaluation of patients with knee osteoarthritis (OA). Yet, there remains a need to assess variability of gait analysis in patients with knee osteoarthritis over time and how pain affects variation in these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service