Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NRCCRMDAG

Domoic acid solution

332 μM in acetonitrile: water (1:19 (v/v))

Synonym(s):

NSC 288031

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO6
CAS Number:
Molecular Weight:
311.33
Beilstein/REAXYS Number:
5126272
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

certified reference material

Quality Level

concentration

332 μM in acetonitrile: water (1:19 (v/v))

storage temp.

2-8°C

InChI

1S/C15H21NO6.3Na/c1-8(4-3-5-9(2)14(19)20)11-7-16-13(15(21)22)10(11)6-12(17)18;;;/h3-5,9-11,13,16H,6-7H2,1-2H3,(H,17,18)(H,19,20)(H,21,22);;;/q;3*+1/p-3/b5-3+,8-4-;;;/t9-,10+,11-,13+;;;/m1.../s1

InChI key

DKZXEHCSMAYDTA-LSCPQTDESA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service